Assay Details
Target Gene Details
Entrez Gene ID: | 17164 |
Gene Name: | MAP kinase-activated protein kinase 2 |
Gene Aliases: |
AA960234, MAPKAP-K2, MK-2, MK2, Rps6kc1 |
Location: |
Chr.1:131053700-131097543 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mapkapk2 | NM_008551.1 | 10 | 1802 | NP_032577.1 |
| AK171912.1 | 1 | 351 | ||
| BC024559.1 | 6 | 1171 | AAH24559.1 | |
| BC063064.1 | 10 | 1802 | AAH63064.1 |
More Information
Set Membership: |
|

Genomic Map