Assay Details
Target Gene Details
Entrez Gene ID: | 66755 |
Gene Name: | RIKEN cDNA 4933415F23 gene |
Gene Aliases: |
- |
Location: |
Chr.1:23100473-23102253 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 4933415F23Rik | NM_025746.2 | 1 | 831 | NP_080022.2 |
| AK005938.1 | 1 | 358 | ||
| AK016819.1 | 1 | 832 | ||
| BC115556.1 | 1 | 832 |
More Information
Set Membership: |
|

Genomic Map