Assay Details
Target Gene Details
Entrez Gene ID: | 241201 |
Gene Name: | cadherin 7, type 2 |
Gene Aliases: |
9330156F07Rik, CDH7L1 |
Location: |
Chr.1:109982327-110141917 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh7 | NM_172853.2 | 12 | 2405 | NP_766441.1 |
| AK034096.1 | 12 | 2405 | BAC28586.1 | |
| AK042657.1 | BAC31323.1 | |||
| AK046426.1 | 5 | 674 | ||
| AK137369.1 | 12 | 2328 | BAE23328.1 | |
| AY267034.1 | 11 | 2260 | AAP94032.1 | |
| BC141119.1 | 11 | 2236 |
More Information
Set Membership: |
|

Genomic Map