Assay Details
Target Gene Details
Entrez Gene ID: | 320272 |
Gene Name: | RIKEN cDNA 9330162B11 gene |
Gene Aliases: |
- |
Location: |
Chr.1:188006576-188008990 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| 9330162B11Rik | NR_038007.1 | 1 | 183 | |
| AK034187.1 | 1 | 184 |
Target Gene Details
Entrez Gene ID: | 26381 |
Gene Name: | estrogen-related receptor gamma |
Gene Aliases: |
ERR3, Errg, NR3B3, mKIAA0832 |
Location: |
Chr.1:187608792-188214884 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Esrrg | NM_001243792.1 | NP_001230721.1 | ||
| NM_011935.3 | NP_036065.1 | |||
| AB093266.1 | BAC41450.1 | |||
| AB221580.1 | ||||
| AF117254.1 | AAD48369.1 | |||
| AK052731.1 | BAC35120.1 | |||
| BC082324.1 | AAH82324.1 | |||
| CJ062694.1 |
More Information
Set Membership: |
|

Genomic Map