Assay Details
Target Gene Details
Entrez Gene ID: | 19250 |
Gene Name: | protein tyrosine phosphatase, non-receptor type 14 |
Gene Aliases: |
C130080N23Rik, OTTMUSG00000022087, PTP36 |
Location: |
Chr.1:189728216-189876695 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptpn14 | NM_008976.2 | 19 | 8493 | NP_033002.2 |
| AK047658.1 | 1 | 416 | ||
| AK081833.1 | 1 | 822 | ||
| AK136544.1 | 1 | 305 | BAE23038.1 |
More Information
Set Membership: |
|

Genomic Map