Assay Details
Target Gene Details
Entrez Gene ID: | 116914 |
Gene Name: | solute carrier family 19 (thiamine transporter), member 2 |
Gene Aliases: |
AV276020, AW322295, DDA1, THTR1, TRMA |
Location: |
Chr.1:164249046-164265385 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc19a2 | NM_001276455.1 | 1 | 213 | NP_001263384.1 |
| NM_054087.3 | 1 | 213 | NP_473428.1 | |
| AF179403.1 | 1 | 204 | AAG43424.1 | |
| AF216204.1 | 1 | 183 | AAK33067.1 | |
| AF326916.1 | AAL85635.1 | |||
| AF360396.1 | 1 | 195 | AAL30451.1 | |
| AF418986.1 | 1 | 204 | AAM47550.1 | |
| AK044825.1 | 1 | 214 | BAC32108.1 | |
| BC034659.1 | 1 | 194 | AAH34659.1 |
More Information
Set Membership: |
|

Genomic Map