Assay Details
Target Gene Details
Entrez Gene ID: | 381319 |
Gene Name: | basic leucine zipper transcription factor, ATF-like 3 |
Gene Aliases: |
9130211I03Rik, Snft |
Location: |
Chr.1:191098414-191108943 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Batf3 | NM_030060.2 | 3 | 633 | NP_084336.1 |
| AK020278.1 | 3 | 634 | BAB32053.1 |
More Information
Set Membership: |
|

Genomic Map