Assay Details
Target Gene Details
Entrez Gene ID: | 228966 |
Gene Name: | protein phosphatase 1, regulatory subunit 3D |
Gene Aliases: |
2610024M03Rik, Gm359 |
Location: |
Chr.2:178411206-178414463 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppp1r3d | NM_001085501.2 | 1 | 1087 | NP_001078970.1 |
More Information
Set Membership: |
|

Genomic Map