Assay Details
Target Gene Details
Entrez Gene ID: | 74194 |
Gene Name: | Rho family GTPase 3 |
Gene Aliases: |
2610017M01Rik, AI661404, Arhe, Rhoe |
Location: |
Chr.2:51130439-51149111 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rnd3 | NM_028810.2 | 5 | 1070 | NP_083086.1 |
| AK011442.1 | 5 | 1068 | BAB27622.1 | |
| AK035195.1 | 5 | 1055 | BAC28975.1 | |
| AK048635.1 | 4 | 2605 | ||
| BC009002.1 | 5 | 1040 | AAH09002.1 |
More Information
Set Membership: |
|

Genomic Map