Assay Details
Target Gene Details
Entrez Gene ID: | 76192 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
1500011G07Rik, 6330583M11Rik, AI431047, AW547313 |
Location: |
Chr.2:150832493-150904745 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd12 | NM_024465.3 | 1 | 247 | NP_077785.2 |
| AK018261.1 | 1 | 179 | ||
| AK160126.1 | 1 | 248 | ||
| BC002138.1 | 1 | 215 | AAH02138.1 |
More Information
Set Membership: |
|

Genomic Map