Assay Details
Target Gene Details
Entrez Gene ID: | 76192 |
Gene Name: | abhydrolase domain containing 12 |
Gene Aliases: |
1500011G07Rik, 6330583M11Rik, AI431047, AW547313 |
Location: |
Chr.2:150832493-150904745 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abhd12 | NM_024465.3 | 13 | 1754 | NP_077785.2 |
| AK160126.1 | 13 | 1758 | ||
| AK178029.1 | 1 | 197 | ||
| AK183821.1 | 1 | 179 | ||
| AK192948.1 | 1 | 149 | ||
| AK194284.1 | 1 | 156 | ||
| AK195364.1 | 1 | 192 | ||
| AK208808.1 | 1 | 197 | ||
| AK211159.1 | 1 | 159 | ||
| AK215521.1 | 1 | 138 | ||
| AK219995.1 | 1 | 178 | ||
| BC002138.1 | 13 | 1723 | AAH02138.1 | |
| BC002263.1 | 9 | 1057 | AAH02263.1 |
More Information
Set Membership: |
|

Genomic Map