Assay Details
Target Gene Details
Entrez Gene ID: | 240916 |
Gene Name: | V-set and immunoglobulin domain containing 8 |
Gene Aliases: |
A030011M19, EG240916 |
Location: |
Chr.1:172555938-172563717 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Vsig8 | NM_177723.4 | 7 | 1569 | NP_808391.2 |
| NR_027644.1 | 6 | 1288 | ||
| AK037228.1 | 7 | 1570 | BAC29767.1 | |
| AW986342.1 | 1 | 246 | ||
| BC064106.1 | 6 | 1288 | AAH64106.1 |
More Information
Set Membership: |
|

Genomic Map