Assay Details
Target Gene Details
Entrez Gene ID: | 226757 |
Gene Name: | WD repeat domain 26 |
Gene Aliases: |
1600024A01Rik, AA693241, AI447817, AU044014, C77982, Gid7 |
Location: |
Chr.1:181173226-181211978 on Build GRCm38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Wdr26 | NM_145514.5 | 14 | 5510 | NP_663489.4 |

Genomic Map