Assay Details
Target Gene Details
Entrez Gene ID: | 11350 |
Gene Name: | c-abl oncogene 1, non-receptor tyrosine kinase |
Gene Aliases: |
AI325092, Abl, E430008G22Rik, c-Abl |
Location: |
Chr.2:31688252-31807093 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abl1 | NM_001112703.2 | 11 | 4350 | NP_001106174.1 |
| NM_001283045.1 | 11 | 3227 | NP_001269974.1 | |
| NM_001283046.1 | 12 | 4555 | NP_001269975.1 | |
| NM_001283047.1 | 12 | 4494 | NP_001269976.1 | |
| NM_009594.4 | 11 | 3121 | NP_033724.2 | |
| AK090095.1 | 11 | 3079 | BAC41088.1 | |
| BC059260.1 | 11 | 3148 | AAH59260.1 | |
| BC103770.1 | 11 | 3574 | AAI03771.1 | |
| BM949741.1 | 1 | 174 | ||
| CA752052.1 | 1 | 84 | ||
| J02995.1 | 11 | 3074 | AAA88241.1 | |
| L10656.1 | 9 | 2675 |

Genomic Map