Assay Details
Target Gene Details
Entrez Gene ID: | 497097 |
Gene Name: | X Kell blood group precursor related family member 4 |
Gene Aliases: |
AY534250, Gm210, XRG4, mKIAA1889 |
Location: |
Chr.1:3199731-3671742 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Xkr4 | NM_001011874.1 | 1 | 797 | NP_001011874.1 |
| AY534250.1 | 1 | 797 | AAT07099.1 |

Genomic Map