Assay Details
Target Gene Details
Entrez Gene ID: | 241452 |
Gene Name: | dehydrogenase/reductase (SDR family) member 9 |
Gene Aliases: |
C730025I08Rik, Rdh15 |
Location: |
Chr.2:69380462-69403086 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhrs9 | NM_175512.2 | 5 | 1279 | NP_780721.1 |
| AF361479.1 | 4 | 1064 | AAN75755.1 | |
| AK050180.1 | 5 | 1297 | BAC34110.1 | |
| AK080914.1 | 5 | 1279 | BAC38075.1 | |
| BC118029.1 | 5 | 1240 |
More Information
Set Membership: |
|

Genomic Map