Assay Details
Target Gene Details
Entrez Gene ID: | 241296 |
Gene Name: | leucine rich repeat containing 8A |
Gene Aliases: |
Lrrc8, mKIAA1437 |
Location: |
Chr.2:30237769-30263790 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lrrc8a | NM_177725.4 | 3 | 1587 | NP_808393.1 |

Genomic Map