Assay Details
Target Gene Details
Entrez Gene ID: | 11432 |
Gene Name: | acid phosphatase 2, lysosomal |
Gene Aliases: |
Acp-2, LAP |
Location: |
Chr.2:91202812-91214098 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acp2 | NM_007387.2 | 11 | 1925 | NP_031413.1 |
| AK080581.1 | 1 | 607 | ||
| AK089401.1 | 11 | 1916 | ||
| AK128963.1 | 11 | 1918 | ||
| AK133756.1 | 11 | 1543 | BAE21825.1 | |
| AK154269.1 | 11 | 1921 | BAE32478.1 | |
| AK170658.1 | 11 | 1926 | ||
| BC023343.1 | 11 | 1907 | AAH23343.1 |
More Information
Set Membership: |
|

Genomic Map