Assay Details
Target Gene Details
Entrez Gene ID: | 60525 |
Gene Name: | acyl-CoA synthetase short-chain family member 2 |
Gene Aliases: |
1110017C11Rik, ACAS, Acas1, Acas2, AceCS1, Acs1 |
Location: |
Chr.2:155517953-155562746 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acss2 | NM_019811.3 | 18 | 2440 | NP_062785.2 |
| AF216873.1 | 18 | 2390 | AAF24510.1 | |
| AK028586.1 | 18 | 2418 | BAC26019.1 | |
| AK029014.1 | 18 | 2408 | BAC26243.1 | |
| AK053877.1 | 18 | 2414 | BAC35571.1 | |
| AK093831.1 | 18 | 2417 | BAC04235.1 | |
| BC051432.1 | 18 | 2440 | AAH51432.1 |
More Information
Set Membership: |
|

Genomic Map