Assay Details
Target Gene Details
Entrez Gene ID: | 66343 |
Gene Name: | transmembrane protein 177 |
Gene Aliases: |
2300008B03Rik |
Location: |
Chr.1:119907890-119913168 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tmem177 | NM_175106.3 | 2 | 585 | NP_780315.1 |
| AK009043.1 | 2 | 591 | ||
| AK076108.1 | 2 | 591 | BAC36188.1 | |
| BC094344.1 | 2 | 585 | AAH94344.1 | |
| BC131993.1 | 1 | 427 | ||
| BC132021.1 | 1 | 427 |

Genomic Map