Assay Details
Target Gene Details
Entrez Gene ID: | 22637 |
Gene Name: | zeta-chain (TCR) associated protein kinase |
Gene Aliases: |
Srk, ZAP-70, mrtle, mur |
Location: |
Chr.1:36761798-36782821 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zap70 | NM_001289765.1 | 2 | 525 | NP_001276694.1 |
| NM_001289766.1 | 2 | 490 | NP_001276695.1 | |
| NM_009539.3 | 2 | 482 | NP_033565.2 | |
| AK039883.1 | 2 | 419 | BAC30471.1 | |
| AK089056.1 | 2 | 455 | ||
| BC029727.1 | 2 | 440 | AAH29727.1 | |
| U04379.1 | 2 | 436 | AAA19250.1 | |
| U77667.1 | 1 | 328 | AAB36538.1 |

Genomic Map