Assay Details
Target Gene Details
Entrez Gene ID: | 75608 |
Gene Name: | charged multivesicular body protein 4B |
Gene Aliases: |
2010012F05Rik, C76846, Snf7-2 |
Location: |
Chr.2:154657026-154694783 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp4b | NM_029362.3 | 1 | 292 | NP_083638.1 |
| AK008205.1 | 1 | 257 | ||
| AK156473.1 | 1 | 257 | BAE33724.1 | |
| AK159193.1 | 1 | 254 | BAE34888.1 | |
| BC006905.1 | AAH06905.1 | |||
| BC011429.1 | 1 | 274 | AAH11429.1 | |
| BC059279.1 | 1 | 254 | AAH59279.1 | |
| CK628208.1 | 1 | 292 |
More Information
Set Membership: |
|

Genomic Map