Assay Details
Target Gene Details
Entrez Gene ID: | 75608 |
Gene Name: | charged multivesicular body protein 4B |
Gene Aliases: |
2010012F05Rik, C76846, Snf7-2 |
Location: |
Chr.2:154657026-154694783 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Chmp4b | NM_029362.3 | 5 | 1318 | NP_083638.1 |
| AK156473.1 | 5 | 1278 | BAE33724.1 | |
| AK159193.1 | 5 | 1280 | BAE34888.1 | |
| BC059279.1 | 5 | 1270 | AAH59279.1 | |
| CF379912.1 | 1 | 321 |
More Information
Set Membership: |
|

Genomic Map