Assay Details
Target Gene Details
Entrez Gene ID: | 380601 |
Gene Name: | FAST kinase domains 5 |
Gene Aliases: |
C78212, mKIAA1792 |
Location: |
Chr.2:130613838-130630027 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fastkd5 | NM_001146084.1 | 3 | 706 | NP_001139556.1 |
| NM_198176.2 | 3 | 743 | NP_937819.2 | |
| AK143929.1 | 3 | 682 | BAE25612.1 | |
| AK220420.1 | 1 | 307 | BAD90466.1 | |
| BB610359.1 | ||||
| BC052898.1 | 3 | 663 | AAH52898.1 |
Target Gene Details
Entrez Gene ID: | 140629 |
Gene Name: | U box domain containing 5 |
Gene Aliases: |
1500010O06Rik, C330018L13Rik, Rnf37, UIP5, Ubce7ip5, Ube7ip5 |
Location: |
Chr.2:130589996-130630038 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ubox5 | NM_001255993.1 | NP_001242922.1 | ||
| NM_001255994.1 | NP_001242923.1 | |||
| NM_080562.5 | NP_542129.2 | |||
| AF360997.1 | AAK51467.1 | |||
| AK005195.1 | ||||
| AK034114.1 | BAC28590.1 | |||
| AK158052.1 | BAE34338.1 | |||
| AK170024.1 | 2 | 599 | ||
| AK170100.1 | ||||
| AK171070.1 |
More Information
Set Membership: |
|

Genomic Map