Assay Details
Target Gene Details
Entrez Gene ID: | 433485 |
Gene Name: | synapse differentiation inducing 1 |
Gene Aliases: |
AW123113, DSPC2, Gm14134, H2C20orf39, OTTMUSG00000015750, Tmem90b |
Location: |
Chr.2:149830053-150004392 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Syndig1 | NM_001085521.2 | 4 | 1446 | NP_001078990.1 |
| BC147352.1 | 3 | 1247 | ||
| BC147353.1 | 3 | 1247 | ||
| BC147660.1 | 3 | 1247 | ||
| BC147661.1 | 3 | 1247 | ||
| BQ176304.1 | 1 | 628 |

Genomic Map