Assay Details
Target Gene Details
Entrez Gene ID: | 16658 |
Gene Name: | v-maf musculoaponeurotic fibrosarcoma oncogene family, protein B (avian) |
Gene Aliases: |
Kreisler, Krml, Krml1, kr |
Location: |
Chr.2:160363677-160367065 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mafb | NM_010658.3 | 1 | 2819 | NP_034788.1 |
| AK143209.1 | 1 | 2821 | BAE25306.1 | |
| AK154830.1 | 1 | 2181 | BAE32860.1 | |
| BC038256.1 | 1 | 2767 | AAH38256.1 |
More Information
Set Membership: |
|

Genomic Map