Assay Details
Target Gene Details
Entrez Gene ID: | 228576 |
Gene Name: | mal, T cell differentiation protein-like |
Gene Aliases: |
BC012256, BENE |
Location: |
Chr.2:127704390-127729897 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mall | NM_145532.3 | 4 | 1202 | NP_663507.1 |
| AK078971.1 | 4 | 1203 | BAC37487.1 |
More Information
Set Membership: |
|

Genomic Map