Assay Details
Target Gene Details
Entrez Gene ID: | 66734 |
Gene Name: | microtubule-associated protein 1 light chain 3 alpha |
Gene Aliases: |
1010001H21Rik, 4922501H04Rik, LC3, LC3a |
Location: |
Chr.2:155276364-155278073 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Map1lc3a | NM_025735.3 | 4 | 836 | NP_080011.1 |
| NR_046364.1 | 3 | 780 | ||
| AK003122.1 | 4 | 747 | BAB22582.1 | |
| AK003716.1 | 4 | 745 | ||
| AK015020.1 | 4 | 755 | ||
| AK219204.1 | 1 | 184 | ||
| BC010596.1 | 4 | 736 | AAH10596.1 | |
| BE993199.1 | 1 | 228 |
More Information
Set Membership: |
|

Genomic Map