Assay Details
Target Gene Details
Entrez Gene ID: | 415115 |
Gene Name: | neuralized-like 2 (Drosophila) |
Gene Aliases: |
Neur2, Ozz, Ozz-E3 |
Location: |
Chr.2:164830730-164833596 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Neurl2 | NM_001082974.2 | 2 | 1106 | NP_001076443.1 |
| AK004524.1 | 2 | 967 | BAB23346.1 | |
| AK180716.1 |
More Information
Set Membership: |
|

Genomic Map