Assay Details
Target Gene Details
Entrez Gene ID: | 226442 |
Gene Name: | zinc finger protein 281 |
Gene Aliases: |
Znf281 |
Location: |
Chr.1:136624901-136630391 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp281 | NM_001160251.1 | 2 | 4416 | NP_001153723.1 |
| NM_177643.4 | 2 | 4412 | NP_808311.1 | |
| AK034110.1 | 1 | 1682 | BAE43287.1 | |
| AK148604.1 | 1 | 1570 |
More Information
Set Membership: |
|

Genomic Map