Assay Details
Target Gene Details
Entrez Gene ID: | 68379 |
Gene Name: | CDKN1A interacting zinc finger protein 1 |
Gene Aliases: |
0610038H21Rik, 2900056O04Rik, LSFR1, ZNF356 |
Location: |
Chr.2:32363006-32378313 on Build GRCm38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ciz1 | NM_001252534.1 | 18 | 2828 | NP_001239463.1 |
| NM_001252536.1 | 19 | 2540 | NP_001239465.1 | |
| NM_001252537.1 | 18 | 2801 | NP_001239466.1 | |
| NM_001252538.1 | 18 | 2491 | NP_001239467.1 | |
| NM_028412.2 | 17 | 2605 | NP_082688.1 | |
| AJ575057.1 | 16 | 2112 | CAE00633.1 | |
| AK013708.1 | 5 | 584 | BAB28962.1 | |
| AK089986.1 | 18 | 2805 | BAC41030.1 | |
| AK137961.1 | 18 | 2407 | BAE23524.1 | |
| AK159103.1 | 18 | 2806 | BAE34819.1 | |
| AK165687.1 | 19 | 2494 | BAE38339.1 | |
| BC018483.1 | 17 | 2569 | AAH18483.1 |

Genomic Map