Assay Details
Target Gene Details
Entrez Gene ID: | 98985 |
Gene Name: | CLP1, cleavage and polyadenylation factor I subunit |
Gene Aliases: |
AI462438, Heab |
Location: |
Chr.2:84723122-84727268 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Clp1 | NM_133840.2 | 2 | 317 | NP_598601.1 |
| AK053169.1 | ||||
| AK077647.1 | 2 | 294 | BAC36924.1 | |
| AK169709.1 | 2 | 326 | BAE41321.1 | |
| BC003237.1 | 2 | 317 | AAH03237.1 |
More Information
Set Membership: |
|

Genomic Map