Assay Details
Target Gene Details
Entrez Gene ID: | 228836 |
Gene Name: | discs, large homolog-associated protein 4 (Drosophila) |
Gene Aliases: |
AI225853, BC024558, DAP-4, DAP4, SAPAP-4, Sapap4, WBP16 |
Location: |
Chr.2:156613633-156764363 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dlgap4 | NM_001042487.1 | 7 | 2489 | NP_001035952.1 |
| NM_001042488.2 | 7 | 1788 | NP_001035953.1 | |
| NM_001277186.1 | 13 | 3724 | NP_001264115.1 | |
| NM_001277187.1 | 12 | 3682 | NP_001264116.1 | |
| NM_146128.6 | 13 | 3733 | NP_666240.4 | |
| AK185553.1 | ||||
| AK218905.1 | 1 | 194 | ||
| BC058948.1 | 7 | 1812 | AAH58948.1 |
More Information
Set Membership: |
|

Genomic Map