Assay Details
Target Gene Details
Entrez Gene ID: | 19213 |
Gene Name: | pancreas specific transcription factor, 1a |
Gene Aliases: |
PTF1-p48, PTF1p48, bHLHa29 |
Location: |
Chr.2:19445663-19447501 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptf1a | NM_018809.2 | 2 | 1060 | NP_061279.2 |
| AB035675.1 | 2 | 1060 | BAA88249.1 | |
| AK007922.2 | 2 | 1086 | BAE43208.1 | |
| BC132505.1 | 2 | 923 | ||
| BC138507.1 | 2 | 923 |
More Information
Set Membership: |
|

Genomic Map