Assay Details
Target Gene Details
Entrez Gene ID: | 11552 |
Gene Name: | adrenergic receptor, alpha 2b |
Gene Aliases: |
Adra-2b, [a]2B, alpha2-C2, alpha2B |
Location: |
Chr.2:127363286-127367221 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adra2b | NM_009633.3 | 1 | 661 | NP_033763.2 |
| AF332049.1 | 1 | 367 | AAK56078.1 | |
| AF332050.1 | 1 | 367 | AAK56079.1 |
More Information
Set Membership: |
|

Genomic Map