Assay Details
Target Gene Details
Entrez Gene ID: | 109778 |
Gene Name: | biliverdin reductase A |
Gene Aliases: |
0610006A11Rik, 2500001N03Rik, Blvr |
Location: |
Chr.2:127070629-127097084 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Blvra | NM_026678.4 | 8 | 884 | NP_080954.4 |
| AK002231.1 | 7 | 823 | BAB21950.1 | |
| AK010847.1 | 8 | 839 | BAB27219.1 | |
| AK172620.1 | 8 | 849 | BAE43098.1 | |
| AK181323.1 | 2 | 237 | ||
| AK182323.1 | 3 | 275 | ||
| BC052146.1 | 8 | 874 | AAH52146.1 | |
| CA983696.1 | 6 | 619 |
More Information
Set Membership: |
|

Genomic Map