Assay Details
Target Gene Details
Entrez Gene ID: | 64685 |
Gene Name: | N-myc (and STAT) interactor |
Gene Aliases: |
- |
Location: |
Chr.2:51948498-51973520 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmi | NM_001141948.1 | 1 | 148 | NP_001135420.1 |
| NM_001141949.1 | 1 | 148 | NP_001135421.1 | |
| NM_019401.2 | 1 | 148 | NP_062274.1 | |
| AF019249.1 | AAB70819.1 | |||
| BY013052.1 | 1 | 148 | ||
| CJ093824.1 | 1 | 144 |
More Information
Set Membership: |
|

Genomic Map