Assay Details
Target Gene Details
Entrez Gene ID: | 64685 |
Gene Name: | N-myc (and STAT) interactor |
Gene Aliases: |
- |
Location: |
Chr.2:51948498-51973520 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nmi | NM_001141948.1 | 8 | 1238 | NP_001135420.1 |
| NM_001141949.1 | 9 | 1278 | NP_001135421.1 | |
| NM_019401.2 | 8 | 1173 | NP_062274.1 | |
| AA217628.1 | 1 | 151 | ||
| AF019249.1 | 8 | 1081 | AAB70819.1 | |
| AK146577.1 | 9 | 1075 | BAE27273.1 | |
| BC002019.1 | 8 | 1033 | AAH02019.1 | |
| CT010361.1 | CAJ18568.1 |
More Information
Set Membership: |
|

Genomic Map