Assay Details
Target Gene Details
Entrez Gene ID: | 99152 |
Gene Name: | anaphase promoting complex subunit 2 |
Gene Aliases: |
9230107K09Rik, AL024279, Apc2, Emi4, Imi4, mKIAA1406 |
Location: |
Chr.2:25272466-25285916 on Build GRCm38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Anapc2 | NM_175300.4 | 13 | 2682 | NP_780509.2 |
| AK033784.1 | 13 | 2650 | BAC28472.1 | |
| AK129351.1 | BAC98161.1 | |||
| AK154465.1 | BAE32605.1 | |||
| AK170676.1 | BAE41951.1 | |||
| AK210019.1 | ||||
| AK210042.1 | ||||
| AK213363.1 | 1 | 142 | ||
| AK215279.1 | 1 | 142 | ||
| BC027351.1 | AAH27351.1 | |||
| BC065421.1 | AAH65421.1 | |||
| BC137583.1 | 13 | 2650 |

Genomic Map