Assay Details
Target Gene Details
Entrez Gene ID: | 14950 |
Gene Name: | histocompatibility 13 |
Gene Aliases: |
1200006O09Rik, 4930443L17Rik, 5031424B04Rik, AV020344, H-13, Hm13, PSL3, Spp |
Location: |
Chr.2:152669437-152708668 on Build GRCm38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| H13 | NM_001159551.1 | NP_001153023.1 | ||
| NM_010376.4 | NP_034506.1 | |||
| AF483214.1 | AAM22075.1 | |||
| AF515662.1 | AAN77098.1 | |||
| AJ345032.1 | CAC87793.1 | |||
| AK007664.1 | BAB25172.1 | |||
| AK012600.1 | ||||
| AK028109.1 | BAC25752.1 | |||
| AK049101.1 | BAC33543.1 | |||
| AK050664.1 | BAC34369.1 | |||
| AK080966.1 | BAC38098.1 | |||
| AK159176.1 | BAE34875.1 | |||
| AK169922.1 | BAE41461.1 | |||
| AK171790.1 | ||||
| DQ168449.1 |
Target Gene Details
Entrez Gene ID: | 66405 |
Gene Name: | malignant T cell amplified sequence 2 |
Gene Aliases: |
2400002F11Rik |
Location: |
Chr.2:152687148-152687946 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcts2 | NM_025543.2 | 1 | 124 | NP_079819.1 |
| AK002563.1 | 1 | 102 | BAB22189.1 | |
| AK010255.1 | 1 | 125 | BAB26798.1 | |
| AK028001.1 | 1 | 103 | BAC25695.1 | |
| BC027507.1 | 1 | 117 | AAH27507.1 |
More Information
Set Membership: |
|

Genomic Map