Assay Details
Target Gene Details
Entrez Gene ID: | 20933 |
Gene Name: | mediator complex subunit 22 |
Gene Aliases: |
AW212655, AW558812, Surf-5, Surf5 |
Location: |
Chr.2:26905262-26910672 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med22 | NM_001033908.1 | 1 | 119 | NP_001029080.1 |
| NM_011513.2 | 1 | 119 | NP_035643.1 | |
| AK144744.1 | 1 | 97 | BAE26044.1 | |
| AK161053.1 | 1 | 60 | BAE36173.1 | |
| AK166250.1 | 1 | 65 | BAE38661.1 | |
| AK166474.1 | 1 | 92 | BAE38795.1 | |
| BC018225.1 | 1 | 109 | AAH18225.1 | |
| BG805496.1 |
More Information
Set Membership: |
|

Genomic Map