Assay Details
Target Gene Details
Entrez Gene ID: | 68975 |
Gene Name: | mediator complex subunit 27 |
Gene Aliases: |
1500015J03Rik, 2310042P07Rik, AA682045, Crsp8, D2Ertd434e |
Location: |
Chr.2:29346810-29524790 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Med27 | NM_001290489.1 | 7 | 774 | NP_001277418.1 |
| NM_026896.5 | 8 | 1048 | NP_081172.2 | |
| AI850753.1 | 1 | 229 | ||
| AK005251.1 | 8 | 1023 | BAB23906.2 | |
| AK088740.1 | 1 | 995 | ||
| BC016537.1 | 8 | 1039 | AAH16537.1 |
More Information
Set Membership: |
|

Genomic Map