Assay Details
Target Gene Details
Entrez Gene ID: | 66404 |
Gene Name: | replication termination factor 2 domain containing 1 |
Gene Aliases: |
1700067C04Rik, 2410001C21Rik, 5730427M17Rik, AA589417 |
Location: |
Chr.2:172440578-172469899 on Build GRCm38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rtfdc1 | NM_025542.2 | 9 | 1490 | NP_079818.1 |
| AK028792.1 | 9 | 1479 | BAC26123.1 | |
| AK053258.1 | 9 | 1503 | BAC35324.1 | |
| AK077823.1 | 9 | 1491 | BAC37024.1 | |
| AK170141.1 | 9 | 1467 | BAE41592.1 | |
| AK182599.1 | ||||
| BC049072.1 | 9 | 1483 | AAH49072.2 | |
| BC096692.1 | 9 | 1439 | AAH96692.1 |

Genomic Map