Assay Details
Target Gene Details
Entrez Gene ID: | 74717 |
Gene Name: | spermatogenesis associated 17 |
Gene Aliases: |
1700065F16Rik, 4930504I07Rik, 4930513F16Rik, MSRG-11 |
Location: |
Chr.1:187009614-187215462 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Spata17 | NM_028848.3 | NP_083124.1 | ||
| NM_029067.1 | 7 | 846 | NP_083343.1 | |
| AK006890.1 | 3 | 431 | BAB24780.1 | |
| AK076836.1 | 3 | 455 | BAC36500.1 | |
| AY747687.1 | 7 | 821 | AAW33969.1 | |
| BC089520.1 | 7 | 821 | AAH89520.1 |

Genomic Map