Assay Details
Target Gene Details
Entrez Gene ID: | 13436 |
Gene Name: | DNA methyltransferase 3B |
Gene Aliases: |
MmuIIIB |
Location: |
Chr.2:153649165-153687730 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnmt3b | NM_001003960.4 | 23 | 3529 | NP_001003960.2 |
| NM_001003961.4 | 24 | 3589 | NP_001003961.2 | |
| NM_001003963.4 | 22 | 3400 | NP_001003963.2 | |
| NM_001122997.2 | 23 | 3477 | NP_001116469.1 | |
| NM_001271744.1 | 23 | 3474 | NP_001258673.1 | |
| NM_001271745.1 | 22 | 3414 | NP_001258674.1 | |
| NM_001271746.1 | 21 | 3285 | NP_001258675.1 | |
| NM_001271747.1 | 20 | 3225 | NP_001258676.1 | |
| NM_010068.5 | 21 | 3340 | NP_034198.3 | |
| AF068626.2 | 24 | 3444 | AAC40178.2 | |
| AF068627.2 | 23 | 3384 | AAC40179.2 | |
| AF068628.2 | 21 | 3195 | AAC40180.2 | |
| AF151969.1 | 24 | 3587 | AAF74515.1 | |
| AF151970.1 | 23 | 3527 | AAF74516.1 | |
| AF151971.1 | 21 | 3338 | AAF74517.1 | |
| AF151972.1 | 22 | 3398 | AAF74518.1 | |
| AF151973.1 | 23 | 3472 | AAF74519.1 | |
| AF151974.1 | 22 | 3412 | AAF74520.1 | |
| AF151975.1 | 21 | 3283 | AAF74521.1 | |
| AF151976.1 | 20 | 3223 | AAF74522.1 | |
| AY078427.1 | 22 | 3255 | AAL85481.1 | |
| BC105677.1 | 23 | 3309 | ||
| BC105922.1 | 23 | 3309 | AAI05923.1 |
More Information
Set Membership: |
|

Genomic Map