Assay Details
Target Gene Details
Entrez Gene ID: | 13482 |
Gene Name: | dipeptidylpeptidase 4 |
Gene Aliases: |
Cd26, Dpp-4, THAM |
Location: |
Chr.2:62330073-62412317 on Build GRCm38 |
Assay Gene Location: | Within Exon 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dpp4 | NM_010074.3 | 26 | 3058 | NP_034204.1 |
| AK085370.1 | 26 | 2641 | BAC39434.1 | |
| AK153939.1 | 26 | 2672 | BAE32266.1 | |
| BC022183.1 | 27 | 2920 | AAH22183.1 | |
| X58384.1 | 26 | 2735 | CAA41274.1 |
More Information
Set Membership: |
|

Genomic Map