Assay Details
Target Gene Details
Entrez Gene ID: | 14239 |
Gene Name: | forkhead box S1 |
Gene Aliases: |
FREAC10, Fkh3, Fkhl18 |
Location: |
Chr.2:152931898-152933208 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Foxs1 | NM_010226.2 | 1 | 1097 | NP_034356.1 |
| AK077935.1 | 1 | 1098 | BAC37074.1 | |
| AK152669.1 | 1 | 1063 | BAE31404.1 | |
| BC096604.1 | 1 | 1064 | AAH96604.1 | |
| BC131937.1 | 1 | 1072 | ||
| BC131939.1 | 1 | 1072 |
More Information
Set Membership: |
|

Genomic Map