Assay Details
Target Gene Details
Entrez Gene ID: | 16492 |
Gene Name: | potassium voltage-gated channel, shaker-related subfamily, member 4 |
Gene Aliases: |
Kv1.4 |
Location: |
Chr.2:107290589-107303587 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kcna4 | NM_021275.4 | 1 | 408 | NP_067250.2 |
| AK036112.1 | 1 | 344 | BAC29309.1 |
More Information
Set Membership: |
|

Genomic Map