Assay Details
Target Gene Details
Entrez Gene ID: | 66044 |
Gene Name: | D-tyrosyl-tRNA deacylase 1 |
Gene Aliases: |
0610006H08Rik, DUE-B, Hars2 |
Location: |
Chr.2:144599953-144768747 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dtd1 | NM_025314.3 | 6 | 1095 | NP_079590.1 |
| AK002246.1 | 6 | 1032 | ||
| AK003594.1 | 6 | 1067 | BAB22882.1 | |
| AK011917.1 | 6 | 1064 | BAB27914.1 | |
| AK158898.1 | 6 | 1152 | BAE34719.1 | |
| BC026537.1 | 6 | 1068 | AAH26537.1 | |
| BE633558.1 | 1 | 160 |
More Information
Set Membership: |
|

Genomic Map