Assay Details
Target Gene Details
Entrez Gene ID: | 100043910 |
Gene Name: | RIKEN cDNA A630075F10 gene |
Gene Aliases: |
4831414D02, Gm4722 |
Location: |
Chr.2:170061240-170070778 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| A630075F10Rik | NR_033632.1 | 1 | 136 | |
| NR_033634.1 | ||||
| AK029198.1 | ||||
| AK042250.1 | 1 | 137 |
Target Gene Details
Entrez Gene ID: | 228911 |
Gene Name: | teashirt zinc finger family member 2 |
Gene Aliases: |
2900073F20Rik, B830045G17, Mtsh2, Sdccag33l, Tsh2, Zfp218, mKIAA4248, teashirt2 |
Location: |
Chr.2:169632496-170071666 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tshz2 | AK046877.1 | 2 | 660 | |
| AK161999.1 | 2 | 990 | BAE36673.1 |
More Information
Set Membership: |
|

Genomic Map